Acid green 50, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Lissamine Green B, Green S

199583-25G
199583-25GEA 64 EUR
199583-25G
Acid green 50, Sigma-Aldrich®
Acid green 50

Lissamine Green B has been used in blood-brain barrier (BBB) studies and is used to study the its effect on herpes simplex virus 1 (HSV-1) corneal infection in animals.

Formel: C₂₇H₂₆N₂O₇S₂
Molecular Weight: 576,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012117
CAS-Nummer: 3087-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be8de2baa4000c4729164a5ff2ed9a33d3dc09b9&vid=19cc12fda401998e7dede7ecae122079eafd6e1b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR