Dimethylethylamine Hydrochloride, Pharmaceutical Analytical Impurities

Lieferant: USP

1A01320
1A01320.EA 530 EUR
1A01320.
Dimethylethylamine Hydrochloride, Pharmaceutical Analytical Impurities
Standards Organische Standards

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestellen

TECHNISCHE DATEN

Bezeichnung Dimethylethylamine Hydrochloride
Synonym(s) N,N-dimethylethanamine, hydrochloride
Environmentally Preferable
CAS Nr. 58114-25-3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR