VWR® RiboReady™ Color Micro RNA Ladder
Lieferant: VWR Chemicals
1B1491-25RXNEA
1040
EUR
1B1491-25RXN
VWR® RiboReady™ Color Micro RNA Ladder
Elektrophorese-Reagenzien
Molekulargewichtsmarker
RiboReady™ Color Micro RNA ladder allows precise sizing of RNA molecules in denaturing acrylamide gels. The apparent sizes of bands in the RiboReady™ Color Micro RNA ladder are 100, 75, 50, 40, 30 and 20 bases in length.
- 6 bands ranging from 20 to 100 bases
- Contains pre-stained nucleic acids that are visible during electrophoresis
Ready to use mixture that does not require heating or addition of a denaturing agent before use.