Sodium triethylborohydride 1 M in THF, AcroSeal®

Lieferant: Thermo Scientific
Gefahr

Synonyme: sodium triethylhydroborate, Borate(1-), triethylhydro-, sodium, (T-4)-, Sodium triethylborohydride

200031000
200031000.EA 108 EUR
200031000.
Sodium triethylborohydride 1 M in THF, AcroSeal®
Sodium triethylborohydride

Appearance: Colorless Solution

Formel: C₆H₁₆BNa
Molecular Weight: 121,99 g/mol
Dichte: 0,89 g/cm³
Flammpunkt: –17 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011704
CAS-Nummer: 17979-81-6
UN: 3399
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Colorless Solution
Titration 0.95 to 1.10 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR