Bromkresolgrün, Pulver pH-Indikator

Lieferant: VWR Chemicals

Synonyme: 3,3',5,5'-Tetrabrom-m-kresolsulfonphthalein, BCG

200125BEA 447 EUR
200125B
Bromkresolgrün, Pulver pH-Indikator
Bromkresolgrün
Formel: C₂₁H₁₄Br₄O₅S
Molecular Weight: 698,02 g/mol
Schmelzpunkt: 217…218 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005874
CAS-Nummer: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Transition range pH 3.8 - 5.4 Yellowish green to blue
Appearance of solution Passes test
Transition range (according to ACS) Passes test
Loss on drying (110°C) Max. 3.00 %
Conforms to ACS Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR