Tetrachrome Stain (MacNeal), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

200190-25G
200190-25GEA 111 EUR
200190-25G
Tetrachrome Stain (MacNeal), Sigma-Aldrich®
Tetrachrome Stain (MacNeal)

Tetrachrome stain along with silver stain is used in MacNeal staining method. Tetrachrome stains nucleus blue and silver stain highlight calcification. Tetrachrome Stain (MacNeal) has been used to stain mouse blood smears to readily identify several morphologically distinct cell types and lineages.

Lagertemperatur: Raumtemperatur
MDL: MFCD00134023
CAS-Nummer: 81142-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR