Azur II eosinate, Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Azure II eosin, Dipotassium 7-(dimethylamino)-N,N-dimethylphenothiazin-3-iminium N,N-dimethyl-7-(methylamino)phenothiazin-3-iminium eosin YS(2-) dichloride, Azure II eosinate

M-Clarity™ Qualitätsstufe = MQ100

200220-100G 200220-25G
200220-100GEA 396 EUR
200220-100G 200220-25G
Azur II eosinate, Pulver, Sigma-Aldrich®
Azur II eosinate

Chief ingredient of Giemsa Stain.

Formel: C₅₁H₄₀Br₄Cl₂N₆Na₂O₅S₂
Lagertemperatur: Room Temperature
MDL: MFCD00084753
CAS-Nummer: 53092-85-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56dc6044a3f48656fb942fede22b70e7a147538c&vid=56dc6044a3f48656fb942fede22b70e7a147538c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength C=0.005 G/L Methanol 645 - 655 nm
Extinction Coefficient ≥49440
Wavelength 243 - 247 nm
Solubility (Turbidity) Hazy to Turbid
Solubility (Color) Dark Blue


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR