Erythrosin B 95 Dye, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: C.I. 45430:2

MQ100

200964-25G 200964-5G
200964-25GEA 238 EUR
200964-25G 200964-5G
Erythrosin B 95 Dye, Sigma-Aldrich®
Erythrosin B

Erythrosin B, also called as eosin B, is a xanthene dye. It belongs to the family of fluorescein dyes. It plays a major role in inhibiting dopamine uptake and high affinity 3H-ouabain binding and ion transport in synaptosomes from rat caudate nucleus. Erythrosin B has been used as a dye in fluorescence lifetime imaging.

Formel: C₂₀H₈I₄O₅
Molecular Weight: 835.89 g/mol
Schmelzpunkt: 303 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00005044
CAS-Nummer: 15905-32-5
EINECS: 240-046-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2400428403cc80d1f8a2edbdc0c236e5fa04e3d&vid=506d9885057163692aef1956066e3fb4444f9e97&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Color): Orange to Red-Orange Conforms to Requirements
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wave Length 258 - 266 nm
Extinction Coefficient ≥13000
Extinction Coefficient C = 0.005g/l, 95% Methanol 5% 0.1M Na2CO3 ≥32000
Carbon 27.3 - 30.2 %
Solubility (Turbidity) 1mg/ml 1M NH4OH Clear
Solubility (Color) Orange to Red


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR