Normal grinding, laboratory kit, NS29
Lieferant: witeg Labortechnik
22.000
201-0210EA 764 EUR
201-0210
Normal grinding, laboratory kit, NS29
Destillationsbauteile Destillations-Bausätze
Supplementary kit for 171021. Consists of the following components:
1 pc Round flask 1000 ml, NS 29/32
1 round flask 1000 ml, 3 necks
2 pcs Capillary 6 mm
1 pc Thermometer 0 + 250 ° C
1 Dest publisher Anschütz-Thiele NS 29/32 - NS 19/26
1 pc Dest column Hempel 300 mm, NS 29/32
1 st Dest column Column Vigreaux 400 mm, NS 29/32
1 evacuation essay NS 29/32
1 Gas Wash Bottle 250 ml, complete