Normal grinding, laboratory kit, NS29
Lieferant: WITEG LABORTECHNIK
21.000
201-0215EA 910 EUR
201-0215
Normal grinding, laboratory kit, NS29
Destillationsbauteile Destillations-Bausätze
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Box with normally ground details for device construction on a macro scale. The kit consists of the following components:
1 round flask, 100 ml, NS 29/32
1 round flask, 250 ml, NS 29/32
1 round flask, 500 ml, NS 29/32
1 st Erlenmeyer flask, 200 ml, NS 29/32
2 capillaries, diam 6 mm
1 radiator, West, length 200 mm, NS-Core and NS-Sleeve 19/26
1 drying tube, NS-Core 19/26
1 drip funnel, cylindrical, graduated, 100 ml, NS-Core 29/32, NS-Sleeve 19/26
6 clamps for NS 19/26
10 clamps for NS 29/32
1 screw cap, GL 18
10 sealing rings, 16 × 6 mm, for GL 18
10 sealing rings, 25 × 8 mm, for GL 25
10 sealing rings, 25 × 12 mm, for GL 25
1 st plug NS 19/26
1 st plug NS 29/32
1 transition piece, NS-Core 29/32, NS-Sleeve 19/26
1 adapter, NS-Core 29/32, ISO thread GL 25
1 distillation set-up, piston sander NS 29/32, ISO thread GL 18, radiator sander NS 19/26
1 drip catcher, piston grinder NS 29/32, radiator grinder NS 19/26
1 distillation set-up, piston sander NS 29/32, sleeve sander NS 29/32
1 thermometer 0 + 250 1/1 ° C
1 trunk tube, radiator grinder NS 19/26
1 evacuation set, bent, with drain pipe and suction pipe radiator grinder NS 19/26, piston grinder NS 29/32
1 stir bar, diam 8 mm
1 stirrer blade, PTFE, 50 × 25 mm
1 pc stirrer sleeve made of polypropylene, with rubber hose