Air/steam inlet tubes, plain, Quickfit®
Lieferant: SciLabware
MF5
201-3059EA 39.6 EUR
201-3059
Air/steam inlet tubes, plain, Quickfit®
Röhrchen Gaseinleitungsrohre
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air/steam inlet tubes are designed to fit the range of Quickfit® thermometer/inlet adapters having size 13 thread. Glass tube with jet.