Soxhlet Extractors, Quickfit®
Lieferant: SciLabware
EX5/32 EX5/43 EX5/53 EX5/55/100 EX5/63 EX5/83 EX5/55
201-8373EA 126 EUR
201-8373 201-8374 201-8375 201-8377 201-8378 201-8380 BIBBEX5/55
Soxhlet Extractors, Quickfit®
Extraktoren Extraktionsapparate, nach Soxhlet
PYREX®, borosilikatglas
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
Standards: Conforms to BS 2071