Condensers, cold finger
Lieferant: SciLabware
FC14/00/SC
201-8389EA 76.9 EUR
201-8389
Condensers, cold finger
Kühler
PYREX® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For immersion into distillation assemblies etc. Can be used to regulate refluxing by adjusting the flow of air or water through it.