Essigsäure (Eisessig) ≥99,9%, AnalaR NORMAPUR® analytical reagent, für die Kryoskopie

Lieferant: VWR Chemicals
Gefahr

20105.292EA 75.3 EUR
20105.292
Essigsäure (Eisessig) ≥99,9%, AnalaR NORMAPUR® analytical reagent, für die Kryoskopie
Essigsäure
Formel: H₃CCOOH
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.9 %
Substances reducing dichromate Passes test ACS
Colouration Max. 10 APHA
Solidification point 16.3 - 16.5 °C
Substances coloured by H2SO4 Max. 150 APHA
Evaporation residue Max. 10 ppm
Ignition residue (SO4) Max. 10 ppm
Water Max. 0.15 %
Cl (Chloride) Max. 1 ppm
NO3 (Nitrate) Max. 1 ppm
SO4 (Sulphate) Max. 1 ppm
Ag (Silver) Max. 0.5 ppm
Al (Aluminium) Max. 0.05 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.01 ppm
Be (Beryllium) Max. 0.01 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.08 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.3 ppm
Ge (Germanium) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.5 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.1 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.1 ppm
Tl (Thallium) Max. 0.05 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.05 ppm
Zr (Zirconium) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR