Dimidiumbromid, GPR RECTAPUR®

Lieferant: VWR Chemicals
Achtung

Synonyme: 3,8-Diamino-5-methyl-6-phenylphenanthridiniumbromid, 2,7-Diamino-10-methyl-9-phenylphenanthridiniumbromid

201272PEA 435 EUR
201272P
Dimidiumbromid, GPR RECTAPUR®
Dimidiumbromid
Formel: C₂₀H₁₈BrN₃
Molecular Weight: 380,29 g/mol
Schmelzpunkt: 243…248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011757
CAS-Nummer: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Dye content (spectrophotometrically) Min. 95 %
Identity (UV/VIS-Spectrum) Passes test
Absorption max. (in methanol) 523 - 528 nm
Spec.Absorptivity (?m;0,0005 %;methanol) 166 - 175
TLC-Test Passes test
Loss on drying (110°C) Max. 5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR