Schnelle rote ITR Base, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Azoic Diazo Component 42, N1,N1-Diethyl-4-methoxymetanilamide, Fast Red ITR, 3-Amino-N,N-diethyl-4-methoxybenzenesulphonamide
201286-100G
201286-100GEA
96.5
EUR
201286-100G
Schnelle rote ITR Base, Sigma-Aldrich®
Fast red ITR base
Fast Red ITR, (Azoic Diazo No. 42)
Formel:
C₁₁H₁₈N₂O₃S Molecular Weight: 258,34 g/mol Schmelzpunkt: 102…105 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009045 CAS-Nummer: 97-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...