(3-Thienyl)-malonsäure 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Propanedioic acid, 3-thienyl-., 3-Thienylmalonic Acid, 3-Thiophenemalonic Acid
201300010
201300010.EA
21.2
EUR
201300010.
(3-Thienyl)-malonsäure 98+%
3-Thiophenemalonic acid
Formel:
C₇H₆O₄S Molecular Weight: 186,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005469 CAS-Nummer: 21080-92-2 EINECS: 244-198-9 |
Spezifikation Testergebnisse
Appearance | beige crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...