Brilliant yellow, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
201375-25G
201375-25GEA
156
EUR
201375-25G
Brilliant yellow, Sigma-Aldrich®
Brilliant yellow
Brilliant Yellow has been used to determine the transfer of inoculum from male moths to females during mating.
Formel:
C₂₆H₂₀N₄O₈S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007489 CAS-Nummer: 3051-11-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...