4-Diazodiphenylamine sulphate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

201464-25G
201464-25GEA 176 EUR
201464-25G
4-Diazodiphenylamine sulphate, Sigma-Aldrich®
4-Diazodiphenylamine sulphate

Variamine Blue RT Salt has been used as a dye in ultraviolet-visible spectrophotometric method based-uricase enzyme for the detection of uric acid in normal urine. It is also used in combination with naphthol AS-MX phosphate-variam to determine the cytochemical visualization of alkaline phosphatase activity.

Formel: C₁₂H₉N₃·H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00011895
CAS-Nummer: 4477-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25ef1a35d0909240c79fd15f9a09311017ac56a0&vid=641148c3d752e02be17bca5e7527a675d2d3d61b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR