Benzoesäure ≥99,7%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

20172.260EA 63 EUR
20172.260
Benzoesäure ≥99,7%, AnalaR NORMAPUR® analytisches Reagens
Benzoesäure
Formel: C₇H₆O₂
Siedepunkt: 249 °C (1013 hPa)
Schmelzpunkt: 121…123 °C
Dichte: 1,316 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002398
CAS-Nummer: 65-85-0
EINECS: 200-618-2
REACH: 01-2119455536-33
Merck Index: 13,01092
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d561f311b11ea8ab2c8f07253e6748e588eb71aa&vid=dcf5e45072cda5106216b497cd29716270ae772c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.7 %
Substances reducing KMnO4 (as O) Passes test
Substances coloured by H2SO4 Passes test
Melting point 121 - 123 °C
Heavy metals (as Pb) Max. 5 ppm
Insolubility in absolute ethanol Max. 50 ppm
Insolubility in diluted ammonia Max. 50 ppm
Insolubility in NaOH 1 N Max. 50 ppm
Ignition residue (SO4) Max. 100 ppm
Total Cl (Chlorine) Max. 50 ppm
Water Max. 0.3 %
SO4 (Sulphate) Max. 30 ppm
Fe (Iron) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR