Eisen(III)-nitrat Nonahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Eisentrinitrat Nonahydrat
2018-01EA
128.03
EUR
2018-01
Eisen(III)-nitrat Nonahydrat, BAKER ANALYZED® ACS, J.T.Baker®
Eisen (III) nitrat Nonahydrat
Formel:
Fe(NO₃)₃·9H₂O Molecular Weight: 404 g/mol Siedepunkt: 125 Schmelzpunkt: 47 Dichte: 1.68 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149708 CAS-Nummer: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1 III Merck Index: 13,04057 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 98.0 - 101.0 % |
Assay (Fe(NO₃)₃9H₂O) (by iodometry) | 98.0 - 101.0 % |
Insoluble Matter | ≤ 0.005 % |
Chloride (Cl) | ≤ 5 ppm |
Sulfate (SO₄) | ≤ 0.01 % |
Calcium (Ca) | ≤ 0.01 % |
Magnesium (Mg) | ≤ 0.005 % |
Potassium (K) | ≤ 0.005 % |
Sodium (Na) | ≤ 0.05 % |
Manganese (Mn) | ≤ 0.01 % |
pH of 5% Solution at 25°C | 1.5 - 2.5 |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...