Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

201820-25G
201820-25GEA 42 EUR
201820-25G
Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®
Acid Red 183

Acid Red 183, Dye content 30 %

Formel: C₁₆H₁₁ClN₄Na₂O₈S₂
Molecular Weight: 532.84 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 6408-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e24db94ce5631aaca9e3be1ce7f5a0680b5f7761&vid=8cdd03ca2be39598d3a6af6b7c19b186622b53e8&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR