Metanilgelb 70% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
M-Clarity™ Qualitätsstufe = MQ200
202029-100G
202029-100GEA
70.2
EUR
202029-100G
Metanilgelb 70% (Farbstoffgehalt), Sigma-Aldrich®
Metanilgelb
Metanil yellow has been used for staining of leukocyte granules. It is also used as a counterstain in microscopic sections.
Metanil yellow is an azo dye. It is used as a counterstain in PAS (periodic acid-Schiff)/iron-hematoxylin stained sections to increase contrast. Metanil yellow changes its color from red at pH 1.2 to yellow at pH 2.3.
Formel:
C₁₈H₁₄N₃NaO₃S Molecular Weight: 375,38 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00007486 CAS-Nummer: 587-98-4 EINECS: 209-608-2 |
Spezifikation Testergebnisse
Appearance (colour) | Gold to Orange |
Appearance (form) | Powder |
pH Transition |
pH 1.2 dark red or pink PH 2.7 yellow |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...