Basic Blue 24 (New Methylene Blue) 90% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
New Methylene Blue N zinc chloride double salt has been used as a photosensitizer for the in vitro photodynamic inactivation of plant-pathogenic fungi Colletotrichum acutatum and Colletotrichum gloeosporioides. It has also been used to determine the in vitro susceptibilities of Trichophyton mentagrophytes and Trichophyton rubrum microconidia to photodynamic antimicrobial chemotherapy (PACT).
Formel:
C₁₈H₂₂ClN₃S·0.5 ZnCl₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078135 CAS-Nummer: 6586-05-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...