Natriumbenzoat, BAKER, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
2021.1000EA
60.36
EUR
2021.1000
2021.9025
Natriumbenzoat, BAKER, Laborreagens, J.T.Baker®
Natriumbenzoat
Formel:
C₆H₅COONa Schmelzpunkt: 410…430 °C Dichte: 1,44 g/cm³ (20 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012463 CAS-Nummer: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Spezifikation Testergebnisse
Assay (dried substance) | 99.0 - 100.5 % |
Alkalinity | passes test |
Heavy Metals (as Pb) | max. 0.001 % |
Identification | passes test |
Organic Volatile Impurities | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...