Tetrakis(triphenylphosphin)palladium(0) 99%

Lieferant: Thermo Fisher Scientific
Achtung

202380010 202380050 202380250 202381000
202380010.EA 68.4 EUR
202380010. 202380050. 202380250. 202381000.
Tetrakis(triphenylphosphin)palladium(0) 99%
Tetrakis(triphenylphosphin)palladium(0)
Formel: C₇₂H₆₀P₄Pd
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00010012
CAS-Nummer: 14221-01-3
EINECS: 238-086-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-green Powder or crystals
Infrared spectrum Conforms
Carbon (C) 73.7 to 76.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR