tert-Butylmethylether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
20256-2.5L
20256-1L
20256-202.7L
20256-2.5LEA
133.88
EUR
20256-2.5L
20256-1L
HONC20256-202.7L
tert-Butylmethylether ≥99,5% (durch GC), Puriss. p.a., Riedel-de Haën™
tert-Butylmethylether
Beilstein Number: 1730942
Formel:
(CH₃)₃COCH₃ Siedepunkt: 55,3 °C (1.013 hPa) Schmelzpunkt: -109 °C Dichte: 0,74 g/cm³ (20 °C) Flammpunkt: -28 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008812 CAS-Nummer: 1634-04-4 EINECS: 216-653-1 UN: 2398 ADR: 3,II Merck Index: 13,06059 |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 99.5 % |
Density (20°C) | 0.736 - 0.744 g/ml |
Refractive index (n 20/D) | 1.362 - 1.376 |
Non-volatile matter | Max. 0.005 % |
Water (Karl Fischer) | Max. 0.05 % |
Free acid (as CH₃COOH) | Max. 0.005 % |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Peroxides (as H₂O₂) | Max. 0.0005 % |
Appearance (color) | complying |
Appearance (form) | complying |
Autoignition Temperature | 705 °F |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...