Salzsäure 25%, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Hydrochloric acid solution, Muriatic acid
20257.296EA
34.6
EUR
20257.296
20257.321
20257.310
20257.401
20257.460
Salzsäure 25%, AnalaR NORMAPUR® analytisches Reagens
Salzsäure
Formel:
HCl Molecular Weight: 36.46 g/mol Schmelzpunkt: –30 °C Dichte: 1.13 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011324 CAS-Nummer: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 II REACH: 01-2119484862-27 |
Spezifikation Testergebnisse
Assay | Min. 25.0 % |
Colouration | Max. 10 APHA |
Free chlorine | Max. 1 ppm |
Ignition residue (SO4) | Max. 5 ppm |
Br (Bromide) | Max. 50 ppm |
NH4 (Ammonium) | Max. 1 ppm |
SO3 (Sulphite) | Max. 1 ppm |
SO4 (Sulphate) | Max. 2 ppm |
Al (Aluminium) | Max. 0.2 ppm |
As (Arsenic) | Max. 0.01 ppm |
Ba (Barium) | Max. 0.05 ppm |
Be (Beryllium) | Max. 0.02 ppm |
Bi (Bismuth) | Max. 0.1 ppm |
Ca (Calcium) | Max. 1 ppm |
Cd (Cadmium) | Max. 0.01 ppm |
Co (Cobalt) | Max. 0.01 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.2 ppm |
Ge (Germanium) | Max. 0.05 ppm |
K (Potassium) | Max. 0.1 ppm |
Li (Lithium) | Max. 0.01 ppm |
Mg (Magnesium) | Max. 0.2 ppm |
Mn (Manganese) | Max. 0.01 ppm |
Mo (Molybdenum) | Max. 0.02 ppm |
Na (Sodium) | Max. 0.5 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.02 ppm |
Sr (Strontium) | Max. 0.01 ppm |
Ti (Titanium) | Max. 0.1 ppm |
Tl (Thallium) | Max. 0.05 ppm |
V (Vanadium) | Max. 0.01 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Zr (Zirconium) | Max. 0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...