Salzsäure 25%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

20257.296EA 34.6 EUR
20257.296 20257.321 20257.310 20257.401 20257.460
Salzsäure 25%, AnalaR NORMAPUR® analytisches Reagens
Salzsäure
Formel: HCl
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: –30 °C
Dichte: 1,13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
REACH: 01-2119484862-27

Bestellen

Spezifikation Testergebnisse

Assay Min. 25.0 %
Colouration Max. 10 APHA
Free chlorine Max. 1 ppm
Ignition residue (SO4) Max. 5 ppm
Br (Bromide) Max. 50 ppm
NH4 (Ammonium) Max. 1 ppm
SO3 (Sulphite) Max. 1 ppm
SO4 (Sulphate) Max. 2 ppm
Al (Aluminium) Max. 0.2 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.05 ppm
Be (Beryllium) Max. 0.02 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.1 ppm
Tl (Thallium) Max. 0.05 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.1 ppm
Zr (Zirconium) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR