D(+)-Biotin, Sigma-Aldrich®
Lieferant: Merck Millipore Calbiochem
Synonyme:
(3aS,4S,6aR)-Biotin, Vitamin H, Vitamin B8, 5-[(3aS,4S,6aR)-2-Oxohexahydro-1H- thieno[3,4-d]imidazol-4-yl]pentansäure, Vitamin B7, Coenzym R
2031-1
2031-1EA
153
EUR
2031-1
D(+)-Biotin, Sigma-Aldrich®
D(+)-Biotin
Formel:
C₁₀H₁₆N₂O₃S Molecular Weight: 244.31 g/mol Schmelzpunkt: 232…233 °C Dichte: 1.41 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005541 CAS-Nummer: 58-85-5 EINECS: 200-399-3 Merck Index: 13,01231 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...