Flußsäure 48%, AnalaR NORMAPUR® zA

Lieferant: VWR Chemicals
Gefahr

20319.291EA 73.9 EUR
20319.291
Flußsäure 48%, AnalaR NORMAPUR® zA
Hydrofluoric acid
Formel: HF
Molecular Weight: 20.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011346
CAS-Nummer: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 (6.1) II

Bestellen

Spezifikation Testergebnisse

Assay Min. 48 %
Colouration Max. 10 APHA
SiF6 (Hexafluorosilicate) Max. 100 ppm
PO4 (Phosphate) Max. 0.5 ppm
SO4 + SO3 (as SO4) Max. 5 ppm
Ag (Silver) Max. 0.02 ppm
Al (Aluminium) Max. 0.1 ppm
As (Arsenic) Max. 0.05 ppm
Ba (Barium) Max. 0.1 ppm
Bi (Bismuth) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Cl (Chloride) Max. 5 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.3 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.02 ppm
Mg (Magnesium) Max. 0.2 ppm
Mn (Manganese) Max. 0.05 ppm
Mo (Molybdenum) Max. 0.05 ppm
Na (Sodium) Max. 0.2 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Ti (Titanium) Max. 0.1 ppm
V (Vanadium) Max. 0.05 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR