Brij® 35 10% in wässriger Lösung, PROTEIN GRADE® Detergens, steril gefiltert, Calbiochem®, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
Brij® L23
203728-50
203728-50EA
277
EUR
203728-50
Brij® 35 10% in wässriger Lösung, PROTEIN GRADE® Detergens, steril gefiltert, Calbiochem®, Millipore®
Brij® 35
Non-ionic detergent useful for the isolation of functional membrane complexes.
BRIJ® 35 detergent be used in protocols that require UV monitoring of the solubilized proteins because of its low UV absorbance. Has been purified to reduce levels of contaminating aldehydes, metals, peroxides, and salts.
Formel:
C₅₈H₁₁₈O₂₄ Molecular Weight: 1199.56 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00080891 CAS-Nummer: 9002-92-0 EINECS: 500-002-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...