Brefeldin A, Millipore®
Lieferant: Merck Millipore Calbiochem
203729-1
203729-1EA
80.4
EUR
203729-1
Brefeldin A, Millipore®
Brefeldin A
Formel:
C₁₆H₂₄O₄ |
MDL:
MFCD00083258 CAS-Nummer: 20350-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...