Tetraethylenglycoldimethylether ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,5,8,11,14-Pentaoxapentadecane
20443.30
20443.A1
20443.30EA
41.3
EUR
20443.30
20443.A1
Tetraethylenglycoldimethylether ≥98%
Tetraethylenglycoldimethylether
Purity: 98+%
CAS No.: 143-24-8
Molecular Formula: CH3(OCH2CH2)4OCH3
Formula Weight: 222.28
Flash Point: 140°(284°F)
Physical Form: Liquid
Density: 1.01
Refractive Index: 1.4330
Melting Point: -30°
Boiling Point: 275-276°
Merck Reference: 14,9208
MDL No.: MFCD00008505
Formel:
C₁₀H₂₂O₅ Siedepunkt: 275…276 °C Schmelzpunkt: –30 °C Dichte: 1,01 Flammpunkt: 140 °C (284 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008505 CAS-Nummer: 143-24-8 EINECS: 205-594-7 Merck Index: 14,9208 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...