Kupfer(II)-2-ethylhexanoat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Ethylcapronsäure Kupfer(II)salz, Kupferbis(2-ethylhexanoat), Heptan-3-carbonsäure Kupfer(II)salz, 2-Ethylhexansäure Kupfer(II)salz
20539.09
20539.18
20539.09EA
74.4
EUR
20539.09
20539.18
Kupfer(II)-2-ethylhexanoat
Kupfer(II)-2-ethylhexanoat
CAS No.: 149-11-1
Molecular Formula: C16H30CuO4
Formula Weight: 349.96
Physical Form: Powder/Lump
Appearance: Blue-green
Melting Point: 252° dec.
MDL No.: MFCD00015695
Formel:
C₁₆H₃₀CuO₄ Molecular Weight: 349.96 g/mol Schmelzpunkt: 252 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015695 CAS-Nummer: 149-11-1 EINECS: 205-731-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...