Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% (durch KMnO₄-Titration), feine Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Ferroammoniumsulfat Hexahydrat, Eisen(II)ammoniumsulfat Hexahydrat, Mohrsches Salz
2054-01EA
205.55
EUR
2054-01
2054-05
2054-02
Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% (durch KMnO₄-Titration), feine Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumeisen(II)sulfat Hexahydrat
Bitte beachten Due to inherent oxidation, ferric ion (Fe³⁺) content may increase on storage.
Formel:
(NH₄)₂Fe(SO₄)₂·6H₂O Schmelzpunkt: 100 °C Dichte: 1,864 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150530 CAS-Nummer: 7783-85-9 EINECS: 233-151-8 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | 98.5 - 101.5% |
For Laboratory,Research,or Manufacturing Use | ≤0.01% |
Assay (Fe(NH₄)₂(SO₄)₂6H₂O) (by KMnO₄ titrn) | 98.5 - 101.5 % |
Insoluble Matter | ≤ 0.01 % |
pH of 5% Solution at 25°C | 3.0 - 5.0 |
Chloride (Cl) | ≤ 0.001 % |
Phosphate (PO₄) | ≤ 0.003 % |
Calcium (Ca) | ≤ 0.005 % |
Copper (Cu) | ≤ 0.003 % |
Ferric Ion (Fe³⁺) | ≤ 0.01 % |
ACS - Magnesium (Mg) | ≤ 0.002 % |
Manganese (Mn) | ≤ 0.01 % |
Potassium (K) | ≤ 0.002 % |
Sodium (Na) | ≤ 0.02 % |
Zinc (Zn) | ≤ 0.003 % |
NOTE: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase | |
on storage. | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...