Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% (durch KMnO₄-Titration), feine Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Ferroammoniumsulfat Hexahydrat, Eisen(II)ammoniumsulfat Hexahydrat, Mohrsches Salz

2054-01EA 205.55 EUR
2054-01 2054-05 2054-02
Ammoniumeisen(II)sulfat Hexahydrat 98,5-101,5% (durch KMnO₄-Titration), feine Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Ammoniumeisen(II)sulfat Hexahydrat

Bitte beachten Due to inherent oxidation, ferric ion (Fe³⁺) content may increase on storage.

Formel: (NH₄)₂Fe(SO₄)₂·6H₂O
Schmelzpunkt: 100 °C
Dichte: 1,864 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150530
CAS-Nummer: 7783-85-9
EINECS: 233-151-8

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 98.5 - 101.5%
For Laboratory,Research,or Manufacturing Use ≤0.01%
Assay (Fe(NH₄)₂(SO₄)₂6H₂O) (by KMnO₄ titrn) 98.5 - 101.5 %
Insoluble Matter ≤ 0.01 %
pH of 5% Solution at 25°C 3.0 - 5.0
Chloride (Cl) ≤ 0.001 %
Phosphate (PO₄) ≤ 0.003 %
Calcium (Ca) ≤ 0.005 %
Copper (Cu) ≤ 0.003 %
Ferric Ion (Fe³⁺) ≤ 0.01 %
ACS - Magnesium (Mg) ≤ 0.002 %
Manganese (Mn) ≤ 0.01 %
Potassium (K) ≤ 0.002 %
Sodium (Na) ≤ 0.02 %
Zinc (Zn) ≤ 0.003 %
NOTE: Due to inherent oxidation, ferric ion (Fe³⁺) content may increase
on storage.
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR