Koffein, Millipore®
Lieferant: Merck Millipore Calbiochem
|
Achtung
|
Synonyme:
Methyltheobromin, 1,3,7-Trimethylxanthin, Guaranin, 1,3,7-Trimethyl-1H,3H-purin-2,6-dion, Coffein, 7-Methyltheobromin, 1,3,7-Trimethyl-2,6(1H,3H)-purindion, Thein, 3,7-Dihydro-1,3,7-trimethyl-1H-purin-2,6-dion, 2,6-Dihydroxy-1,3,7-trimethylpurin, 1,3,7-Trimethyl-3,7-dihydro-2H-purin-2,6-dion, Teein, 1,3,7-Trimethyl-3,7-dihydro-1H-purin-2,6-dion
205548-500
205548-500EA
143
EUR
205548-500
Koffein, Millipore®
Koffein
Formel:
C₈H₁₀N₄O₂ Molecular Weight: 194.19 g/mol Schmelzpunkt: 235…239 °C Dichte: 1.23 g/cm³ (19 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005758 CAS-Nummer: 58-08-2 EINECS: 200-362-1 Merck Index: 13,01636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...