Phenol 99,0-100,5%, einzelne Kristalle Ph. Eur.

Lieferant: VWR Chemicals
Gefahr

20596.297EA 177 EUR
20596.297 20596.468
Phenol 99,0-100,5%, einzelne Kristalle Ph. Eur.
Phenol
Formel: C₆H₅OH
Siedepunkt: 181,8 °C (1013 hPa)
Dichte: 1,06 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002143
CAS-Nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Identification C Passes test
Appearance of solution Passes test
Acidity Passes test
Freezing point Min. 39.5 °C
Residue on evaporation Max. 0.05 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR