Phenol 99,0-100,5%, einzelne Kristalle, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Phenylalkohol, Hydroxybenzol, Carbolsäure
20599.231EA
63.3
EUR
20599.231
20599.297
20599.260
Phenol 99,0-100,5%, einzelne Kristalle, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Phenol
Formel:
C₆H₅OH Molecular Weight: 94.11 g/mol Siedepunkt: 181.8 °C (1013 hPa) Schmelzpunkt: 40.8 °C Dichte: 1.06 g/cm³ (20 °C) Flammpunkt: 81 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002143 CAS-Nummer: 108-95-2 EINECS: 203-632-7 UN: 1671 ADR: 6.1 II REACH: 01-2119471329-32 Merck Index: 12,07390 |
Spezifikation Testergebnisse
Assay | 99.0 - 100.5 % |
Assay (GC) | Min. 99.5 % |
Acidity | Passes test Ph.Eur. |
Appearance of solution | Passes test Ph.Eur. |
Clarity of solution | Passes test ACS |
Identification A | Passes test Ph.Eur. |
Identification B | Passes test Ph.Eur. |
Identification C | Passes test Ph.Eur. |
Freezing point | Min. 40.5 °C |
o-Cresol | Max. 0.05 % |
m-Cresol | Max. 0.05 % |
p-Cresol | Max. 0.05 % |
Evaporation residue | Max. 100 ppm |
Heavy metals (as Pb) | Max. 10 ppm |
Insolubility in water | Max. 50 ppm |
Water | Max. 0.2 % |
Cl (Chloride) | Max. 5 ppm |
Cu (Copper) | Max. 0.1 ppm |
Fe (Iron) | Max. 1 ppm |
K (Potassium) | Max. 10 ppm |
Na (Sodium) | Max. 10 ppm |
Pb (Lead) | Max. 0.1 ppm |
Zn (Zinc) | Max. 0.5 ppm |
Conforms to ACS | Passes test |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...