Phenol 99,0-100,5%, einzelne Kristalle, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Phenylalkohol, Hydroxybenzol, Carbolsäure

20599.231EA 63.3 EUR
20599.231 20599.297 20599.260
Phenol 99,0-100,5%, einzelne Kristalle, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
Phenol
Formel: C₆H₅OH
Siedepunkt: 181,8 °C (1013 hPa)
Schmelzpunkt: 40,8 °C
Dichte: 1,06 g/cm³ (20 °C)
Flammpunkt: 81 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002143
CAS-Nummer: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 100.5 %
Assay (GC) Min. 99.5 %
Acidity Passes test Ph.Eur.
Appearance of solution Passes test Ph.Eur.
Clarity of solution Passes test ACS
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
Freezing point Min. 40.5 °C
o-Cresol Max. 0.05 %
m-Cresol Max. 0.05 %
p-Cresol Max. 0.05 %
Evaporation residue Max. 100 ppm
Heavy metals (as Pb) Max. 10 ppm
Insolubility in water Max. 50 ppm
Water Max. 0.2 %
Cl (Chloride) Max. 5 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 1 ppm
K (Potassium) Max. 10 ppm
Na (Sodium) Max. 10 ppm
Pb (Lead) Max. 0.1 ppm
Zn (Zinc) Max. 0.5 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR