Molekularsieb A5 (0,5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Lieferant: Merck
Synonyme:
Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
20619
20619.EA
271
EUR
20619.
Molekularsieb A5 (0,5 nm, 5 Å) Moisture Trap volume 200 cc, 1/8 in. fitting, Supelco®
Molekularsieb A5 (0,5 nm, 5 Å)
MDL:
MFCD00131613 CAS-Nummer: 69912-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...