Agar, Pulver für die Bakteriologie

Lieferant: VWR Chemicals

20767.232EA 184 EUR
20767.232 20767.298
Agar, Pulver für die Bakteriologie
Agar
Schmelzpunkt: 90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Clarity after autoclaving (*) Max. 15 NTU
Clarity before autoclaving Max. 15 NTU
Gelling point (1.5 %; water) (*) 30 - 40 °C
Gel strength (1.5 %; water) (*) 700 - 1000 g/cm²
pH after autoclaving (1.5 %) (*) 6.0 - 7.5
pH before autoclaving (1.5 %) 6.0 - 7.5
Ignition residue (SO4) Max. 5 %
As (Arsenic) Max. 5 ppm
Pb (Lead) Max. 2 ppm
(*) tested on a 1.5% solution after autoclaving (121 °C) and cooled to 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR