L(+)-Glutaminsäure 98,5-101,5 USP, multi-compendial, J.T.Baker®

Lieferant: Avantor

Synonyme: (S)-(+)-1-Aminopropan-1,3-dicarbonsäure, (S)-(+)-2-Aminoglutarsäure, L(+)-1-Aminopropan-1,3-dicarbonsäure, (S)-(+)-α-Aminoglutarsäure, (S)-(+)-Glutaminsäure, L(+)-2-Amino-1,5-pentandisäure, (S)-(+)-2-Amino-1,5-pentandisäure, L(+)-α-Aminoglutarsäure, H-Glu-OH, L(+)-2-Aminoglutarsäure

2077-06EA 186.6 EUR
2077-06
L(+)-Glutaminsäure 98,5-101,5 USP, multi-compendial, J.T.Baker®
L(+)-Glutaminsäure
Formel: HO₂CCH₂CH₂CH(NH₂)CO₂H
Molecular Weight: 147.13 g/mol
Siedepunkt: 325
Schmelzpunkt: 160
Dichte: 1.538 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002634
CAS-Nummer: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1cb2faefcc53483f0b3bfccd6c12b611e487241&vid=a462989da97f7186f34fef7de9cad6596c2589ee&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Assay (C₅H₉NO₄) (dried basis) 98.5 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 30.5 - 32.5 Degree
EP/BP - Ninhydrin-Positive Substances Passes Test
EP/BP - Chloride (Cl) ≤200 ppm
EP/BP - Sulfate (SO₄) ≤300 ppm
EP/BP - Ammonium (NH₄) ≤200 ppm
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Drying ≤0.5 %
EP/BP - Ash (sulfated) ≤0.1 %
USP - Assay (C₅H₉NO₄) (dried basis) 98.5 - 101.5 %
USP - Residue on Ignition ≤0.1 %
USP - Chloride (Cl) ≤0.02 %
USP - Sulfate (SO₄) ≤0.02 %
USP - Iron (Fe) ≤10 µg/g
USP - Related Compounds - Individual Impurities ≤0.5 %
USP - Related Compounds - Total Impurities ≤2.0 %
USP - Specific Rotation [Å]²º^D (+) 31.5 - 32.5 Degree
USP - Loss on Drying at 105°C ≤0.1 %
USP - Identification Passes Test
Endotoxin Concentration, EU/g ≤3.0
No Class 1,2,3 or other solvents are used or produced in the ≤ 0.1 %
manufacturing or purification of the product. Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR