1-Pentanol ≥98,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: 1-Pentylalkohol, n-Amylalkohol, Butylcarbinol, 1-Hydroxypentan

20791.292EA 64.4 EUR
20791.292
1-Pentanol ≥98,5%, AnalaR NORMAPUR® analytisches Reagens
1-Pentanol
Formel: H₃C(CH₂)₃CH₂OH
Molecular Weight: 88,15 g/mol
Siedepunkt: 137,9 °C (1013 hPa)
Schmelzpunkt: –79 °C
Dichte: 0,81 g/cm³ (20 °C)
Flammpunkt: 48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002977
CAS-Nummer: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 98.5 %
Boiling point 137 - 139 °C
Density (20/4) 0.813 - 0.817
Non-volatile residue Max. 50 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR