1-Butanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: 1-Butylalkohol, 1-Hydroxybutan, n-Butanol, Propylcarbinol

20810.298EA 72 EUR
20810.298 20810.323 20810.460
1-Butanol ≥99,5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
1-Butanol
Formel: H₃C(CH₂)₂CH₂OH
Siedepunkt: 117,6 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0,812 g/cm³ (20 °C)
Flammpunkt: 30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002964
CAS-Nummer: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Acidity or alkalinity Max. 0.0005 meq/g
Boiling point 117 - 119 °C
Colouration Max. 10 APHA
Density (20/4) 0.807 - 0.812
Density (20/20) 0.808 - 0.813
Aldehydes + ketones (as C3H7CHO) Max. 0.01 %
Butan-2-ol Max. 0.05 %
Dibutyl ether Max. 0.2 %
Evaporation residue Max. 10 ppm
Water Max. 0.1 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR