Coated filter, ORBO™ 80
Lieferant: Merck
20811
20811.EA 0 EUR
20811.
Coated filter, ORBO™ 80
Filter Glasfaserfilter
ORBO™ 80 is a glass fiber filter coated with 1-(2-pyridyl)piperazine.
  • No cassettes
It is used for sampling diisocyanates in air.