Ethanol 96% (v/v), AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

20823.293EA 35.5 EUR
20823.293 20823.327 20823.362 20823.555 20823.460 20823.290 20823.930
Ethanol 96% (v/v), AnalaR NORMAPUR® analytisches Reagens
Ethanol

Bitte beachten Wichtiger Hinweis: Diese Chemikalie unterliegt regulatorischen Bestimmungen. Bitte wenden Sie sich an Ihr lokales Vertriebszentrum.

Formel: C₂H₅OH
Siedepunkt: 78,3 °C (1013 hPa)
Schmelzpunkt: –117 °C
Dichte: 0,7895 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.8 %
Assay (V/V)
IR Spectrum Passes test
UV Spectrum Passes test
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 78.0 - 78.5 °C
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 20 APHA
Aldehydes (as CH3CHO) Max. 5 ppm
Aldehydes + ketones (as CH3CHO) Max. 10 ppm
Benzene Max. 5 ppm
Evaporation residue Max. 5 ppm
Formaldehyde Max. 1 ppm
Furaldehyde Max. 1 ppm
Ketones (as CH3COCH3) Max. 10 ppm
Methanol Max. 0.02 %
Propan-2-ol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 4 ppm
Total heavy alcohols Max. 100 ppm
Water 5.2 - 8.2 %
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR