2-Propanol ≥99,0%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol
20839.297EA
29.8
EUR
20839.297
20839.366
20839.468
20839.550
20839.930
2-Propanol ≥99,0%, GPR RECTAPUR®
2-Propanol
Formel:
(CH₃)₂CHOH Molecular Weight: 60.1 g/mol Siedepunkt: 82 Schmelzpunkt: –89 °C Dichte: 0.786 g/cm³ (20 °C) Flammpunkt: 12 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011674 CAS-Nummer: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3 II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Spezifikation Testergebnisse
Assay | Min. 99.0 % |
Acidity | Passes test |
Acidity or alkalinity | Passes test |
Benzene and related substances | Passes test |
IR Spectrum | Passes test |
Peroxides | Passes test |
Boiling point | 82.2 - 82.6 °C |
Density (20/20) | 0.785 - 0.789 |
Density (25/25) | 0.783 - 0.790 |
Brechungsindex (20°C) | 1.376 - 1.378 |
Evaporation residue | Max. 20 ppm |
Water | Max. 0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...