2-Propanol ≥99,7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol

20842.298EA 47.9 EUR
20842.298 20842.323 20842.312 20842.367 20842.460 20842.550 20842.330 20842.930
2-Propanol ≥99,7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytisches Reagens
2-Propanol
Formel: (CH₃)₂CHOH
Siedepunkt: 82 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0,786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Gehalt (ber. auf wasserfreie Substanz) Min. 99.7 %
Benzene and related substances Passes test
IR-Spektrum Passes test
Peroxides Passes test
Solubility in water Entspricht ACS
Acidity Max. 0.0001 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 81,5 - 82,6 °C
Colouration Max. 10 APHA
Density (20/4) 0.784 - 0.786
Density (20/20) 0.785 - 0.787
Density (25/25) 0.783 - 0.790
Brechungsindex (20°C) 1.376 - 1.378
Substances coloured by H2SO4 Max. 60 APHA
Aldehyde (als CH3CHO) Max. 2 ppm
Carbonyl-Verbindungen (als Aceton) Max. 20 ppm
Carbonyl-Verbindungen (als Propionaldehyd) Max. 20 ppm
Ethanol Max. 100 ppm
Evaporation residue Max. 10 ppm
Formaldehyde Max. 2 ppm
Methanol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 5 ppm
Water Max. 0.1 %
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.2 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.05 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR